Specifications
- Antibody Type:Primary
- Antigen Name:tumor necrosis factor (ligand) superfamily, member 10
- Antigen Symbol:TNFSF10
- Clonality:Monoclonal
- Clone:M66-4B
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Lyophilized
- Gene ID:P50591
- Antigen Synonyms:TNFSF10
- Storage Temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping Temperature:RT
- Immunogen:The monoclonal antibody was produced from a hybridoma using purified recombinant, insect cell derived, hTGFβ1.
- Purification:This IgG1k antibody was purified on a Protein G affinity column.
- Cat. No.:10813-676
- Supplier no.:XP-5729-M
Specifications
About this item
ELISA: In a sandwich ELISA (assuming 100 uL/well), a concentration of 4.0-8.0 ug/mL of this antibody will detect at least 100 pg/mL of recombinant human TGFbeta1 when used in conjunction with compatible secondary reagents. Western Blot: To detect hTGFbeta1 by Western Blot analysis this antibody can be used at a concentration of 2.0-4.0 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hTGFbeta1 is 2.0-4.0 ng/lane, under non-reducing conditions. In a dot blot, a 1:500 dilution of this antibody exhibited complete cross reactivity with human TGFbeta2 and TGFbeta3 isotypes. Reduced cross reactivity was shown at a 1:1000 dilution.
Type: Primary
Antigen: TNFSF10
Clonality: Monoclonal
Clone: M66-4B
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human