Anti-TNFSF13B Mouse Monoclonal Antibody [clone: 2B]
Catalog # 10813-674
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:tumor necrosis factor (ligand) superfamily, member 13b
- Antigen Symbol:TNFSF13B
- Clonality:Monoclonal
- Clone:2B
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Lyophilized
- Gene ID:Q9Y275
- Antigen Synonyms:TNFSF13B
- Storage Temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping Temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c mice using recombinant human BAFF as the immunizing antigen.
- Purification:This IgG1/K antibody was purified from ascites fluid on a Protein A chromatography column.
- Cat. No.:10813-674
- Supplier no.:XP-5728-M
Specifications
About this item
ELISA: In a sandwich ELISA (assuming 100 uL/well), a concentration of 5.0-6.0 ug/mL of this antibody will detect at least 1000 pg/mL of recombinant human BAFF when used with our biotinylated antigen affinity purified anti-human BAFF (XP-5105Bt) as the detection antibody at a concentration of approximately 0.25-0.50 ug/mL. Western Blot: To detect hBAFF by Western Blot analysis this antibody can be used at a concentration of 0.20-0.40 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hBAFF is 2.0-4.0 ng/lane, under non-reducing conditions.
Type: Primary
Antigen: TNFSF13B
Clonality: Monoclonal
Clone: 2B
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human