- Antibody Type:Primary
- Antigen Name:chemokine (C-C motif) ligand 25
- Antigen Symbol:CCL25
- Clonality:Monoclonal
- Clone:1.2_4G1-1G4-1C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Neutralisation:Yes
- Form:Lyophilized
- Gene ID:O15444
- Antigen Synonyms:CCL25
- Storage Temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping Temperature:RT
- Immunogen:The monoclonal antibody was produced in Balb/c X ICR F1 mice using recombinant hTECK as the immunizing antigen.
- Purification:This IgG1k antibody was purified from ascites fluid on a Protein A affinity column.
- Cat. No.:10813-670
- Supplier no.:XP-5725-M
This gene belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. TECK displays chemotactic activity for dendritic cells, thymocytes, and activated macrophages but is inactive on peripheral blood lymphocytes and neutrophils. TECK binds to chemokine receptor CCR9. Two alternatively spliced variants encoding two different isoforms have been identified.
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of hTECK (100 ng/mL), a concentration of 10.0-20.0 ug/mL of this antibody is required. ELISA: In a sandwich ELISA (assuming 100 uL/well), a concentration of 1.0-2.0 ug/mL of this antibody will detect at least 300 pg/mL of recombinant human TECK when used with our biotinylated antigen affinity purified anti-human TECK (XP-5280Bt) as the detection antibody at a concentration of approximately 0.5-1.0 ug/mL. Western Blot: To detect hTECK by Western Blot analysis this antibody can be used at a concentration of 0.25-0.50 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hTECK is 0.50-1.0 ng/lane, under non-reducing conditions.
Type: Primary
Antigen: CCL25
Clonality: Monoclonal
Clone: 1.2_4G1-1G4-1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human