To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CCR5 (phospho Ser349) Antibody
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P51681
- Antigen Synonyms:CKR5|CMKBR5|C-C CKR-5|CHEMR13|CCR-5
- Modification:Ser 349
- Storage Buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular Weight:40 kDa
- Storage Temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:CCR5 (phospho Ser349) antibody was raised against a peptide sequence around phosphorylation site of Serine 349 (E-I-S (p)-V-G) derived from Human CCR5.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. No.:10813-904
- Supplier no.:79-782
Receptor for a number of inflammatory CC-chemokines including MIP-1-alpha, MIP-1-beta and RANTES and subsequently transduces a signal by increasing the intracellular calcium ion level. May play a role in the control of granulocytic lineage proliferation or differentiation. Acts as a coreceptor (CD4 being the primary receptor) for HIV-1 R5 isolates.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: CCR5 (phospho Ser349) Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human