OxiSelect™ AOPP Assay Kit, Cell Biolabs
Catalog # 102964-896
Supplier: Cell Biolabs
CAS Number:
Specifications
- Description:AOPP Assay
- Size:200 Assays
- Storage Temperature:Room Temperature, 4°C and –20°C
- Cat. No.:102964-896
- Supplier no.:STA-318
Specifications
About this item
OxiSelect™ AOPP Assay Kit provides a convenient method to assess oxidative stress
Advanced Oxidation Protein Products (AOPP) are uremic toxins created during oxidative stress through the reaction of plasma proteins with chlorinated oxidants such as chloramines or hypochlorous acid. AOPP levels are elevated in patients with renal complications, diabetes mellitus, and atherosclerosis, as well as HIV-positive patients.