About this item
AAV-3 Helper Free Expression Complete Systems contain the following constructs needed to package your recombinant serotype-3 adeno-associated virus: an expression vector, pHelper plasmid, a serotype-specific Rep-Cap plasmid, and an optional GFP expression vector for making a control virus
AAV Helper Free Expression Complete Systems are available for native serotypes 1 through 6. The shRNA expression vectors contain a U6 promoter. All other expression vectors contain a CMV promoter which is optimal for general gene expression, with the exception of our promoterless vector which is useful if you wish to clone in a gene-specific promoter along with your gene of interest.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Format:Purified Plasmid
- Storage Temperature:–20°C