About this item
RAPAd® Adenoviral Expression Systems allow you to package recombinant adenovirus in only 2-3 weeks, compared to 2-3 months with other methods, while significantly reducing the population of replication-competent virus
Expression systems for gene expression studies are available with shuttle vectors containing either a CMV or RSV promoter. If your gene of interest is better expressed with a different promoter, choose our Promoterless Adenoviral Expression System where you can clone in the optimal promoter along with your gene. Additional systems are available specifically for introduction of miRNA and shRNA sequences.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Format:Purified Plasmid
- Storage Temperature:–20°C