Order Entry
United States
ContactUsLinkComponent
 
Conical Tip for the Pomeroy Ear Syringe, OR Grade, Sklar
  10198-358
 :  Sklar
undefined
Conical Tip for the Pomeroy Ear Syringe, OR Grade, Sklar
  10198-358
 :  Sklar
 :  67-3411
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    Pomeroy Ear Syringe, Conical Tip, Straight, 2"
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Cat. No.:
    10198-358
  • Supplier no.:
    67-3411

 

 

Sklar's Conical Tip for the pomeroy ear syringe is used for irrigation and cleaning of the ear canal.

The conical, or cone shaped, tip of 0.25 mm and it will enter the ear canal further than a bulbous tip. The size and shape of the patient's ear, along with user preference, will determine which type of tip is utilized. Tips are connected to the canister of the syringe either by screwing on or with a luer Lock, which also provides a seal and prevents fluid from leaking. This product utilizes a screw-on connection.