ContactUsLinkComponent
Pomeroy Ear Syringe Shield, OR Grade, Sklar
Pomeroy Ear Syringe Shield, OR Grade, Sklar
  10198-356
 :  Sklar
undefined
Pomeroy Ear Syringe Shield, OR Grade, Sklar
  10198-356
 :  Sklar
 :  67-3410
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Pomeroy Ear Syringe Chrome Plated Shield Only, Non-Luer Lock
  • Material:
    Chrome plated
  • Environmentally Preferable:
  • Cat. No.:
    10198-356
  • Supplier no.:
    67-3410
Sklar's Pomeroy Ear Syringe Shield is a concave shield used between the canister and the tip to deflect any solution as it flushes out of the ear.
The shield is connected to the canister either by a screw-on tip or with a Luer Lock, which also provides a seal and prevents fluid from leaking. This product is a shield for use with non Luer Lock connecting tips.