About this item
ViraSafe™ Ecotropic Lentiviral Expression Systems contain an expression vector plus all the packaging plasmids necessary to make recombinant lentivirus
Ecotropic lentiviruses are constructed with an envelope protein that can only readily infect mouse or rat cells. If you do not plan to infect cells of any other species, making an ecotropic lentivirus can add an additional level of safety since it will not easily infect human cells. However, ecotropic viruses are typically less stable than VSVG-pseudotyped (pantropic) viruses.
The lentiviral expression systems listed here vary only by the structure of the expression vector. The shRNA systems contain a U6-containing expression vector. All other systems contain an expression vector with an EF-1 promoter, with the exception of our Promoterless Expression System. These systems are designed for experiments where a gene-specific promoter is desired; simply clone the promoter and gene of interest together into the vector.
Specifications
- Format:Purified Plasmid
- Storage Temperature:–20°C