Glucose Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
Cell Biolabs' Glucose Assay Kits measure total glucose present in food or biological samples
Glucose Oxidase first oxidizes glucose, generating hydrogen peroxide that is detected by a probe. The signal produced correlates to the level of glucose in the sample. Glucose levels in an unknown sample are calculated based on a glucose standard curve. Kits are available with either colorimetric or fluorometric detection in a 96-well plate format.
Specifications
- Storage Temperature:Room Temperature and –20°C
- Sensitivity:6.25 µM (colorimetric), 1.56 µM (fluorometric)