Hydroxyproline Assay Kit, Cell Biolabs
Hydroxyproline Assay Kit, 96 Assays, Colorimetric
Catalog # 102966-250
Marketsource
Supplier: Cell Biolabs
Supplier #: STA-675
Hydroxyproline Assay Kit, Cell Biolabs
Catalog # 102966-250
Supplier: Cell Biolabs
Marketsource
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Cell Biolabs' Hydroxyproline Assay Kit is a quantitative colorimetric assay for measuring the hydroxyproline concentration in protein samples. Hydroxyproline in acid hydrolyzed samples is converted to pyrrole through a reaction with Chlormine T, which then reacts with Ehrlich’s Reagent to form a chromophore that is detected at 540-560nm
Hydroxyproline levels in samples are calculated based on a hydroxyproline standard curve. Detect hydroxyproline levels as low as 50 µM.
Documents
STA-675
Manual
Specifications
Specifications
- Description:Hydroxyproline Assay Kit
- Size:96 Assays
- Method:Colorimetric
- Cat. No.:102966-250
- Sensitivity:50 µM
- Storage Temperature:4°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



