Order Entry
United States
ContactUsLinkComponent
 
CytoSelect™ WST-1 Cell Proliferation Assay Reagent, Cell Biolabs
  102963-852
 :  
undefined
CytoSelect™ WST-1 Cell Proliferation Assay Reagent, Cell Biolabs
  102963-852
 :  CBA-253
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Source:
  • Species:
  • Description:
    WST-1 Cell Proliferation Assay Reagent
  • Size:
    10 mL
  • Environmentally Preferable:
  • Storage Temperature:
    4 °C (Short Term) or –20 °C (Long Term) Protected from Light
  • Cat. No.:
    102963-852
  • Assay:
    Colorimetric
  • Supplier no.:
    CBA-253

 

 

Cell Biolabs’ CytoSelect™ WST-1 Cell Proliferation Assay Reagent provides a colorimetric format for measuring and monitoring cell proliferation.  The 10 mL volume is sufficient for the evaluation of 960 assays in ten 96-well plates or 192 assays in eight 24-well plates.  Cells can be plated and then treated with compounds or agents that affect proliferation.  Cells are then detected with the proliferation reagent, which is converted in live cells from WST-1 to the formazan form in the presence of cellular NADH and an electron mediator.

An increase in cell proliferation is accompanied by increased signal, while a decrease in cell proliferation (and signal) can indicate the toxic effects of compounds or suboptimal culture conditions.  The assay principles are basic and can be applied to most eukaryotic cell lines, including adherent and non-adherent cells and certain tissues.  This cell proliferation reagent can be used to detect proliferation in bacteria, yeast, fungi, protozoa as well as cultured mammalian and piscine cells.