- Description:MTT Cell Proliferation Assay
- Size:960 Assays
- Environmentally Preferable:
- Storage Temperature:–20°C Protected from Light
- Cat. No.:102963-850
- Method:Colorimetric
- Supplier no.:CBA-252
Cell Biolabs’ CytoSelect™ MTT Cell Proliferation Assay provides a colorimetric format for measuring and monitoring cell proliferation. Cells can be plated and then treated with compounds or agents that affect proliferation. Cells are then detected with the proliferation reagent, which is converted in live cells from the yellow tetrazole MTT to the purple formazan form by a cellular reductase (Figure 1). An increase in cell proliferation is accompanied by an increased signal, while a decrease in cell proliferation (and signal) can indicate the toxic effects of compounds or suboptimal culture conditions. The assay principles are basic and can be applied to most eukaryotic cell lines, including adherent and non-adherent cells and certain tissues. This cell proliferation reagent can be used to detect proliferation in bacteria, yeast, fungi, protozoa as well as cultured mammalian and piscine cells
The kit contains sufficient reagents for the evaluation of 960 assays in 96-well plates or 192 assays in 24-well plates.