Cell Biolabs' ViraDuctin™ Retrovirus Transduction Kit provides superior retroviral transduction efficiencies in a variety of cell lines, even when compared to transductions in the presence of Polybrene®
This system is ideal for many primary cells as well as immobilized cells. The kit uses a proprietary reagent cocktail that forms supercomplex with retrovirus. The result is an increase in infection efficiency of 2- to 6-fold. The kit is espeically useful for transduction of non-permissive cells, including many primary cells and stem cells.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Environmentally Preferable: