Gag-Pol Retroviral Vector, Cell Biolabs
102965-758
:
- Description:pCMV-Gag-Pol
- Size:10 μg
- Format:Purified Plasmid
- Storage Temperature:–20°C
- Cat. No.:102965-758
- Supplier no.:RV-111
pCMV-Gag-Pol Retroviral Vector contains both gag and pol genes from MMLV-based retrovirus
This vector may be co-transfected with other retroviral constructs to make recombinant retroviral particles.