96-well Cellular Senescence Assay (SA β-Gal Activity), Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
Cell Biolabs' 96-Well Cellular Senescence Activity Assay provides an efficient method to measure Senescence Associated (SA) ß-galactosidase activity
SA-ß-Gal catalyzes the hydrolysis of X-gal, which produces a blue color in senescent cells. Quantify senescence using a fluorescence plate reader.
Specifications
- Storage Temperature:4°C and –20°C