Global DNA Methylation ELISA Kit (5’-methyl-2’-deoxycytidine Quantitation), Cell Biolabs
The Global DNA Methylation ELISA Kit is a competitive ELISA for the quantitative measurement of 5-methylcytosine (5MedCyd)
The unknown 5MedCyd samples or 5MedCyd standards are first added to a 5MedCyd DNA conjugate coated EIA plate. After a brief incubation, an anti-5MedCyd monoclonal antibody is added, followed by an HRP conjugated secondary antibody. The 5MedCyd content in unknown samples is determined by comparison with a predetermined 5MedCyd standard curve. This kit is suitable for use with genomic DNA from cells or tissues, and can detect as little as 150 nM of 5MedCyd.
- Assay Type (ELISA with LOV):Competitive
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:150 nM