ContactUsLinkComponent
Tissue-Tek® FormaGO® Formalin Absorption Granules, Sakura® Finetek
Tissue-Tek® FormaGO® Formalin Absorption Granules, Sakura® Finetek
Catalog # 10852-358
Supplier:  Sakura Finetek
undefined
Tissue-Tek® FormaGO® Formalin Absorption Granules, Sakura® Finetek
Catalog # 10852-358
Supplier:  Sakura Finetek
Supplier Number:  9158
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    FormaGO® Formalin Absorption Granules
  • Size:
    2/pack
  • Cat. No.:
    10852-358
  • Packaging:
    Jar
  • Supplier no.:
    9158
Specifications
About this item
Effective neutralizing agent for accidental formalin spills
Granules retain their form making it easy to pick up with a broom and brush, while leaving less residue after cleaning the spill.