OxiSelect™ ORAC Activity Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
The Oxygen Radical Antioxidant Capacity (ORAC) assay is a powerful tool to measure the antioxidant capacity of biomolecules
OxiSelect™ ORAC Activity Assay measures this capacity in less than 2 hours from a wide variety of sample types. The assay is suitable for use with plasma, cell fractions, and tissue lysates, as well as solid and aqueous nutrition samples. Obtain results in about 2 hours.
Specifications
- Storage Temperature:Room Temperature, 4°C and –20°C