Order Entry
United States
ContactUsLinkComponent
 
GFP Quantitation Kit, Cell Biolabs
Catalog # 102963-620
Supplier:  Cell Biolabs
CAS Number:  
undefined
GFP Quantitation Kit, Cell Biolabs
Catalog # 102963-620
Supplier:  Cell Biolabs
Supplier Number:  AKR-120
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    GFP Quantitation Kit
  • Size:
    100 Assays
  • Environmentally Preferable:
  • Storage Temperature:
    4°C
  • Sensitivity:
    100 ng/mL
  • Cat. No.:
    102963-620
  • Supplier no.:
    AKR-120

Specifications

About this item

The GFP Quantitation Kit provides a convenient plate-based alternative to measuring GFP fluorescence by FACS analysis

The quantity of GFP in a sample is determined by comparing its fluorescence reading with that of known recombinant GFP standard curve. A proprietary GFP quench solution is also included for determining autofluorescence of cell or tissue sample. The kit has detection sensitivity limit of 100 ng/mL.