Order Entry
Puerto Rico
ContactUsLinkComponent
GFP Retroviral Vectors, Cell Biolabs
GFP Retroviral Vectors, Cell Biolabs
Supplier:  Cell Biolabs
Certificates
undefined
GFP Retroviral Vectors, Cell Biolabs
Supplier:  Cell Biolabs

Specifications

  • Format:
    Purified Plasmid
  • Storage Temperature:
    –20°C

Specifications

Related Information