Order Entry
Puerto Rico
ContactUsLinkComponent
pMZs Retroviral Expression Vector, Cell Biolabs
pMZs Retroviral Expression Vector, Cell Biolabs
Catalog # 102966-176
Supplier:  Cell Biolabs
CAS Number:  
undefined
pMZs Retroviral Expression Vector, Cell Biolabs
Catalog # 102966-176
Supplier:  Cell Biolabs
Supplier Number:  RTV-030
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    pMZs
  • Size:
    10 µg
  • Format:
    Purified Plasmid
  • Storage Temperature:
    –20°C
  • Cat. No.:
    102966-176
  • Supplier no.:
    RTV-030

Specifications

About this item

Cell Biolabs' pMZs vector is suitable for making retrovirus for general gene expression

Clone your gene of interest into one of these vectors and then transfect the construct into a retroviral packaging cell line, 293T cells, or 293RTV cells.