pMYs Retroviral Expression Vectors, Cell Biolabs
Cell Biolabs' pMYs vectors are suitable for making retrovirus for gene expression in hematopoietic stem cells
Clone your gene of interest into one of these vectors and then transfect the construct into a retroviral packaging cell line, 293T cells, or 293RTV cells.
- Format:Purified Plasmid
- Storage Temperature:–20°C