Tissue-Tek® Quick-Ray™ Microarray System, Sakura
Product Details & Documents
Tissue Micro Arrays (TMA) are a collection of multiple tissue cores arranged in an XY fashion inside a paraffin block.
- Collection of multiple tissues
- Hallowed tip to remove cores
- Pre-formed paraffin recipient blocks
The Quick-Ray technique uses a hallowed tip to remove cores of 1, 2, 3, 5 mm from tissue which is embedded in paraffin (donor block). The extracted tissue is then inserted into pre-formed paraffin recipient blocks.
Kit is complete with extractor, replaceable punch tips in 1, 2, 3 and 5 mm size; recipient blocks of 1, 2, 3 and 5 mm size; a 1 mm punch guide; recipient base mold.
Each recipient block creates 300-400 tissue sections at 4-5 microns. Kit comes with complete instructions for use with packaging.
Specifications
- Description:Tissue-Tek® Quick-Ray™ Microarray System
Accessories
Product Information
- Catalog #AvailabilityPrice
- Recipient Block 2 mm core. Replaceable pre-formed paraffin block, Single wrapped blockRecipient Block 2 mm core. Replaceable pre-formed paraffin block, Single wrapped block89199-870Log in for availabilityEach - $144.00
Product Information
- Catalog #AvailabilityPrice
- Recipient Block 1 mm core. Replaceable pre-formed paraffin block, Single wrapped blockRecipient Block 1 mm core. Replaceable pre-formed paraffin block, Single wrapped block89199-868Log in for availabilityEach - $136.00
Product Information
- Catalog #AvailabilityPrice
- Tissue Puncher Tip, 5 mm. Replaceable tip for extractor, 1/packageTissue Puncher Tip, 5 mm. Replaceable tip for extractor, 1/package89199-866Log in for availabilityEach - $933.00
Product Information
- Catalog #AvailabilityPrice
- Tissue Puncher Tip, 3 mm. Replaceable tip for extractor, 1/packageTissue Puncher Tip, 3 mm. Replaceable tip for extractor, 1/package89199-864Log in for availabilityEach - $968.00
Product Information
- Catalog #AvailabilityPrice
- Tissue Puncher Tip, 2 mm. Replaceable tip for extractor, 1/packageTissue Puncher Tip, 2 mm. Replaceable tip for extractor, 1/package89199-862Log in for availabilityEach - $968.00
Product Information
- Catalog #AvailabilityPrice
- Quick-Ray Base Mold. Base molds to fit recipient blocks, 3 in a packageQuick-Ray Base Mold. Base molds to fit recipient blocks, 3 in a package89199-876Log in for availabilityCase of 3 - $56.70
Product Information
- Catalog #AvailabilityPrice
- Recipient Block 5 mm core. Replaceable pre-formed paraffin block, Single wrapped blockRecipient Block 5 mm core. Replaceable pre-formed paraffin block, Single wrapped block89199-874Log in for availabilityEach - $138.00
Product Information
- Catalog #AvailabilityPrice
- Recipient Block 3 mm core. Replaceable pre-formed paraffin block, Single wrapped blockRecipient Block 3 mm core. Replaceable pre-formed paraffin block, Single wrapped block89199-872Log in for availabilityEach - $144.00
Product Information
- Catalog #AvailabilityPrice
- Tissue Puncher Tip, 1 mm. Replaceable tip for extractor, 1/packageTissue Puncher Tip, 1 mm. Replaceable tip for extractor, 1/package89199-860Log in for availabilityEach - $968.00
Product Information
- Catalog #AvailabilityPrice
- Tissue Puncher Tip Guide, 1 mm. Helps align the 1 mm tip with the recipient block, 1/packageTissue Puncher Tip Guide, 1 mm. Helps align the 1 mm tip with the recipient block, 1/package89199-858Log in for availabilityEach - $682.00