Anti-METAP1 Rabbit Polyclonal Antibody
Catalog # 89518-488
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Methionyl Aminopeptidase 1
- Antigen Symbol:METAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:Center
- Gene ID:23173
- Antigen Synonyms:MetAP1A|MAP1A
- Amino Acid Number:188 to 216
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:43.215 kDa
- Sequence:HGIPDRRPLQEGDIC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.48mg/ml
- Shipping Temperature:4 °C
- Immunogen:This METAP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 188-216 amino acids from the Central region of human METAP1.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89518-488
- Supplier no.:AP17161C
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: METAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human