To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Source:Bacillus polymyxa
- Size:1 g
- Enzyme Name:Dipase
- Tested Applications:Restriction Enzyme
- Cat. No.:10630-956
- Supplier no.:P5318
Dispase is a mild protease which cleaves fibronectin, collagen IV and to a lesser extent collagen I.
Lyophilized and partially purified dispase is used for Neutral Protease (Dispase) Assay, tissue disaggregation and subcultivation, prevention of unwanted cell clumping, preparation of cells for culture, separation of intact epidermis from dermis and intact epithelial sheet in culture from the substratum, harvest and transfer of cell lines, gentle and intact detachment of epidermal cells.