Order Entry
Puerto Rico
ContactUsLinkComponent
Clamp Holder, 19 mm Grip, Chemglass
Clamp Holder, 19 mm Grip, Chemglass
Catalog # 80063-652
Supplier:  Chemglass
Small Business Enterprise
undefined
Clamp Holder, 19 mm Grip, Chemglass
Catalog # 80063-652
Supplier:  Chemglass
Supplier Number:  CG-9250-01
Small Business Enterprise

Specifications

  • Description:
    Clamp Holder
  • Cat. No.:
    80063-652
  • Supplier no.:
    CG-9250-01

Specifications

About this item

Nickel-plated zinc alloy clamp holder has high tensile strength and corrosion resistance.

  • Useful wherever clamping at 90° is required
  • Large thumb screws allow easy adjusting