About this item
Brilliant Violet 510™ anti-human CD49d [9F10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cynomolgus, Dog (Canine), Horse (Equine), Rhesus, Sheep (Ovine), Squirrel Monkey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: 9F10
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cat (Feline);Cattle (Bovine;Cow);Chimpanzee;Common Marmoset;Cynomolgus;Dog (Canine);Horse (Equine);Rhesus;Sheep (Ovine);Squirrel Monkey
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10757-848
- 10021-020
- Antigen Symbol:
- CD49d
- CD49d
- Antigen Synonyms:
- Integrin α4 chain|α4 integrin|VLA-4 α chain|ITGA4
- Integrin α4 chain|α4 integrin|VLA-4 α chain
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 510
- Brilliant Violet® 510
- Clone:
- 9F10
- 9F10
- Reactivity:
- Squirrel monkey, Cat, Baboon, Horse, African green monkey, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human, Bovine, Dog
- Squirrel monkey, Cat, Baboon, Horse, African green monkey, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human, Bovine, Dog
- Host:
- Mouse
- Mouse
- Gene ID:
- 3676
- 3676
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 510™ and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 510™ and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT