Anti-PRKAG3 Rabbit Polyclonal Antibody
89518-794
: Abgent
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 3
- Antigen Symbol:PRKAG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:53632
- Antigen Synonyms:AMPKG3|AMPK gamma3|gamma-3 subunit|AMPK gamma-3 chain|5'-AMP-activated protein kinase
- Amino Acid Number:426 to 454
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:54.258 kDa
- Sequence:GEVIDRIAREQVHRC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This PRKAG3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 426-454 amino acids from the C-terminal region of human PRKAG3.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89518-794
- Supplier no.:AP16887B
Western Blot: 1:1000
Type: Primary
Antigen: PRKAG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human