Anti-MBD2 Rabbit Polyclonal Antibody
400 µL
Catalog # 89514-214
Supplier: Abgent
Supplier #: AP19883C
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Western Blot: 1:1000, Immunofluorescence: 1:10~50,
Type: Primary
Antigen: MBD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Size:400 µL
- Clonality:Polyclonal
- Molecular Weight:43.255 kDa
- Purification:Protein A & Peptide affinity purification
- Epitope:Center
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Host:Rabbit
- Sequence:NKVKSDPQRMNEQPRC
- Concentration:0.29mg/ml
- Cat. No.:89514-214
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:MBD2
- Gene ID:8932
- Immunogen:This MBD2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 250-279 amino acids from the Central region of human MBD2.
- Amino Acid Number:250 to 279
- Conjugation:Unconjugated
- Antigen Synonyms:MBD2a|NY-CO-41|DMTase
- Antigen Name:Methyl-CpG Binding Domain Protein 2
- ELISA:Yes
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



