Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-TRAM1L1 Rabbit Polyclonal Antibody
  89517-176
 :  Abgent
Anti-TRAM1L1 Rabbit Polyclonal Antibody
  89517-176
 :  Abgent
 :  AP18041B

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Translocation Associated Membrane Protein 1-like 1
  • Antigen Symbol:
    TRAM1L1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    C-Terminal
  • Gene ID:
    133022
  • Amino Acid Number:
    325 to 353
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    42.162 kDa
  • Sequence:
    RSSKKRTENGVGVEC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This TRAM1L1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 325-353 amino acids from the C-terminal region of human TRAM1L1.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89517-176
  • Supplier no.:
    AP18041B

 

 

Western Blot: 1:1000

Type: Primary
Antigen: TRAM1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human