Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-BEX4 Rabbit Polyclonal Antibody
Anti-BEX4 Rabbit Polyclonal Antibody
Catalog # 89517-450
Supplier:  Abgent
Anti-BEX4 Rabbit Polyclonal Antibody
Catalog # 89517-450
Supplier:  Abgent
Supplier Number:  AP17957C

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Brain Expressed X Linked 4
  • Antigen Symbol:
    BEX4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    Center
  • Gene ID:
    56271
  • Antigen Synonyms:
    NADE3|BEX4|FLJ10097|BEXL1
  • Amino Acid Number:
    54 to 82
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    14.067 kDa
  • Sequence:
    RHIEHNEARDDVERC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This BEX4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 54-82 amino acids from the Central region of human BEX4.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89517-450
  • Supplier no.:
    AP17957C

Specifications

About this item

Western Blot: 1:1000

Type: Primary
Antigen: BEX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human