Anti-TAAR9 Rabbit Polyclonal Antibody
Catalog # 89516-346
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Trace Amine-associated Receptor 9
- Antigen Symbol:TAAR9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:134860
- Antigen Synonyms:TRAR3|TA3|TAR3|TAR9
- Amino Acid Number:229 to 255
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:39.016 kDa
- Sequence:SYKERVAKRERKC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This TAAR9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 229-255 amino acids from the C-terminal region of human TAAR9.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89516-346
- Supplier no.:AP18516B
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: TAAR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse