Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD31 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: WM59]
Anti-CD31 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: WM59]
Supplier:  Biolegend
Certificates
Anti-CD31 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: WM59]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 10759-462
  • 10757-768
  • Antigen Symbol:
  • CD31
  • CD31
  • Antigen Synonyms:
  • PECAM-1|EndoCAM|PECAM1
  • PECAM-1|EndoCAM|PECAM1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 421
  • Brilliant Violet® 421
  • Clone:
  • WM59
  • WM59
  • Reactivity:
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5175
  • 5175
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications