Anti-CD3 Mouse Monoclonal Antibody [clone: HIT3a]
Supplier: Biolegend
Certificates
About this item
Ultra-LEAF™ Purified anti-human CD3 [HIT3a]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, IP, Activ, IHC; Size: 1 mg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: HIT3a
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Specifications
- Catalog No:
- 300331-BL
- 300332-BL
- Antigen Symbol:
- CD3
- CD3
- Antigen Synonyms:
- T3|CD3E|CD3ε
- T3|CD3E|CD3ε
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HIT3a
- HIT3a
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 916
- 916
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Form:
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Purification:
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Concentration:
- 1.00 mg/mL
- 1.00 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C. This Ultra-LEAF™ solution contains no preservative; handle under aseptic conditions.
- The antibody solution should be stored undiluted between 2 °C and 8 °C. This Ultra-LEAF™ solution contains no preservative; handle under aseptic conditions.
- Shipping Temperature:
- Ambient RT
- Ambient RT