ContactUsLinkComponent
Dual Probe Digital Thermometer with Alarm, Thermco
Dual Probe Digital Thermometer with Alarm, Thermco
Catalog # 10025-442
Supplier:  Thermco
undefined
Dual Probe Digital Thermometer with Alarm, Thermco
Catalog # 10025-442
Supplier:  Thermco
Supplier Number:  ACC8100MATB
Specifications
  • Description:
    Dual Probe Digital Thermometer
  • Cat. No.:
    10025-442
  • Probe:
    Dual probe
  • Range:
    −50...70 °C (−58...158 °F)
  • Supplier no.:
    ACC8100MATB
Specifications
About this item
Dual temperature thermometer to monitor freezer and refrigerator simultaneously, with min/max time and date stamp alarm.
  • Audible and visual alarms
  • 5mL vaccine bottle probes filled with bio-safe medium