Rtx®-OPPesticides2 Columns (Fused Silica), Restek
Supplier: Restek
Certificates
About this item
Using sophisticated computer modeling software, Restek has created two stationary phases for separating the 53 organophosphorus pesticides (OPP) listed in EPA method 8141.
- Application-specific columns for organophosphorus pesticides; best column combination for U.S. EPA method 8141
- Low bleed-ideal for GC-FPD, GC-NPD, or GC-MS analyses
- Stable to 330 to 340 °C
- Proprietary Crossbond® phases
Separation is improved and analysis time is significantly reduced, compared to other columns. The extended upper temperature limit of these phases (330 °C) allows analysts to bake out high molecular weight contamination typically associated with pesticide samples. The low-bleed columns are a perfect match for sensitive detection systems.
Specifications
- Catalog No:
- 10850-188
- 10850-186
- 10850-184
- 10850-182
- Description:
- Rtx-OPPesticides2
- Rtx-OPPesticides2
- Rtx-OPPesticides2
- Rtx-OPPesticides2
- Temperature Limit:
- -20 to 310/330 °C
- -20 to 310/330 °C
- -20 to 300/330 °C
- -20 to 310/330 °C