Intermediate-Polarity Deactivated MXT® Guard/Retention Gap Columns/Transfer Lines (passivated stainless steel), Restek
Supplier: Restek
Certificates
About this item
Intermediate-polarity deactivated MXT® guard/retention gap columns are useful for a wide range of applications and are proven to be compatible with most common solvents.
- Useful for a wide range of applications
- Compatible with most common solvents
- Maximum temperature: 325 °C
- Sold in 5-meter lengths
Constructed from passivated stainless steel.