Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-NKP30 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-S30]
Anti-NKP30 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-S30]
Catalog # 102995-858
Supplier:  Cell Sciences
Anti-NKP30 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-S30]
Catalog # 102995-858
Supplier:  Cell Sciences
Supplier Number:  CDM324

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    NKP30
  • Clonality:
    Monoclonal
  • Clone:
    B-S30
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Antigen Synonyms:
    CD337|1C7|LY117|NCR3|NKp30|MALS
  • Cat. No.:
    102995-858
  • Supplier no.:
    CDM324

Specifications

About this item

Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: NKP30
Clonality: Monoclonal
Clone: B-S30
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human