Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MICB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-D54]
Anti-MICB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-D54]
Catalog # 102995-832
Supplier:  Cell Sciences
Anti-MICB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-D54]
Catalog # 102995-832
Supplier:  Cell Sciences
Supplier Number:  CDM311

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    MHC Class I Polypeptide-Related Sequence B
  • Antigen Symbol:
    MICB
  • Clonality:
    Monoclonal
  • Clone:
    B-D54
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Antigen Synonyms:
    MHC Class I Chain-Related Protein B|MHC Class I Mic-B Antigen|MHC Class I-Like Molecule PERB11.2-IMX|PERB11.2
  • Cat. No.:
    102995-832
  • Supplier no.:
    CDM311

Specifications

About this item

Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: MICB
Clonality: Monoclonal
Clone: B-D54
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human