Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-IL6 Mouse Monoclonal Antibody [clone: B-F6]
  102995-760
Anti-IL6 Mouse Monoclonal Antibody [clone: B-F6]
  102995-760
 :  CDM267

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Interleukin 6
  • Antigen Symbol:
    IL6
  • Clonality:
    Monoclonal
  • Clone:
    B-F6
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    500 µg
  • Antigen Synonyms:
    HSF|HGF|IFNB2|interferon beta 2|BSF2|IL-6
  • Cat. No.:
    102995-760
  • Supplier no.:
    CDM267

 

 

No effect on IL-6 induced proliferation on B9 cell line

Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: B-F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human