Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 1 Beta
- Antigen Symbol:IL1B
- Clonality:Monoclonal
- Clone:B-A15
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:500 µg
- Antigen Synonyms:catabolin|IL-1 beta|preinterleukin 1 beta|IL1 B|pro-interleukin-1-beta|IL1-BETA|interleukin-1 beta|IL-1|IL1B|IL1F2
- Cat. No.:102995-754
- Supplier no.:CDM259
Specifications
About this item
Blocks IL-1 beta induced proliferation on D10S murine cell line. Suitable for intra-cytoplasmic flow cytometry.
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: B-A15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human