Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:CD40 ligand
- Antigen Symbol:CD40L
- Clonality:Monoclonal
- Clone:B-B29
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:200 µg
- Antigen Synonyms:hCD40L|TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
- Cat. No.:102995-640
- Supplier no.:CDM183
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: B-B29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: