Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Interleukin 17 A
- Antigen Symbol:IL17A
- Clonality:Monoclonal
- Clone:B-B51
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Size:500 µg
- Antigen Synonyms:IL-17|IL17|CTLA8|IL-17A|CTLA-8|Cytotoxic T-lymphocyte-associated serine esterase 8
- Cat. No.:102995-600
- Supplier no.:CDM114
Neutralizes the bioactivity of IL-17A on keratinocytes cell line HaCaT (measurement of beta defensin-2 production)
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: B-B51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human