Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:TNF-Related Apoptosis-Inducing Ligand
- Antigen Symbol:TRAIL
- Clonality:Monoclonal
- Clone:B-S23
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:200 µg
- Antigen Synonyms:CD253 antigen|TRAIL protein|Tumor necrosis factor ligand|Apo-2 Ligand|superfamily member 10|TNF-Related Apoptosis-Inducing Ligand|TRAIL|CD253|TL2|Apo-2L ,Apo2L|APO2L|TNFSF10
- Cat. No.:102995-546
- Supplier no.:CDM076
Specifications
About this item
Enhances TRAIL induced apoptosis. Recognizes the soluble antigen.
Type: Primary
Antigen: TRAIL
Clonality: Monoclonal
Clone: B-S23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human