Order Entry
Puerto Rico
ContactUsLinkComponent
Kay Aorta Clamp, OR Grade, Sklar
Kay Aorta Clamp, OR Grade, Sklar
Catalog # 10196-288
Supplier:  Sklar
undefined
Kay Aorta Clamp, OR Grade, Sklar
Catalog # 10196-288
Supplier:  Sklar
Supplier Number:  52-6775

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Kay Aorta Clamp, Double Angled
  • Length:
    20.3 cm (8")
  • Material:
    Stainless steel
  • Cat. No.:
    10196-288
  • Supplier no.:
    52-6775

Specifications

About this item

Kay Aorta Clamps are constructed of stainless steel

Industry standard.