- Description:Weitlaner Retractor
- Length:14.0 cm (5½")
- Material:Stainless steel
- Tip configuration:Solid Blades
- Cat. No.:10195-230
- Supplier no.:22-8760
Sklar's Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site
It has curved shanks that lead to blades, most commonly including between two and six, sharp or blunt, outward curving prongs. The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries. It is also useful with small, deep incisions and soft tissue dissection at a superficial level.