Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Catalog # 60775-881
Supplier: Micro Essential
Specifications
- Description:pH Paper, Mikro Hydrion Dispenser Double Roll (1-12)
- Cat. No.:60775-881
- Supplier no.:M-1200
Specifications
About this item
Hydrion® Double Roll pH Paper set the standard for quality, accuracy, and reliability
- Composed of Special Wet Strength Paper
- Color chart provides clear color matches based on spectral color changes for sample comparison
- These Mikro paper rolls measure 180 x 5/32 inches
These rolls test the 1.0-12.0 pH range with each full pH unit showing a distinct color variation.
Packaging: Packaged in a dual roll protective plastic dispenser and contained in a polypropylene case to guard against sunlight and laboratory hazards.