ContactUsLinkComponent
Moisture Dish, Humboldt
Moisture Dish, Humboldt
Catalog # 72410-006
Small Business Enterprise
Moisture Dish, Humboldt
Catalog # 72410-006
Supplier Number:  H-17150
Small Business Enterprise
Specifications
  • Environmentally Preferable:
  • Cat. No.:
    72410-006
  • I.D.×D:
    57×19 mm (2¼×¾")
  • Supplier no.:
    H-17150
Specifications
About this item
Aluminum moisture dish with inverted slip-in cover and beaded edge for tight sealing, can be handled with crucible tongs.